missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VASP [p Ser157] Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33584-20ul
This item is not returnable.
View return policy
Description
VASP Monoclonal antibody specifically detects VASP in Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| VASP | |
| Monoclonal | |
| Western Blot 1:2000 - 1:5000, ELISA Recommended starting concentration is 1 μg/mL | |
| vasodilator-stimulated phosphoprotein | |
| A synthetic phosphorylated peptide around S157 of human VASP (NP_003361.1).,, Sequence:, RRVSNA | |
| 20 μL | |
| Phospho Specific | |
| 7408 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction