missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Vasorin/SLIT-like 2 Antibody Pair [HRP]

Product Code. 18689060 Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
This item is not returnable. View return policy

Product Code. 18689060

Brand: Novus Biologicals™ NBP2794185Plates

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Vasorin/SLIT-like 2.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 23.4 pg/mL
Assay Sensitivity 23.4-1500 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Vasorin/SLIT-like 2.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias Protein slit-like 2, SLITL2, slit-like 2, slit-like 2 (Drosophila), vasorin
Gene ID (Entrez) 114990
Gene Symbol VASN
Host Species Rabbit
Kit Contents
  • Standard: Each vial contains 46 ng of recombinant Human Vasorin/SLIT-like 2.
  • Capture Antibody: 0.5 mg/mL of rabbit anti-Human Vasorin/SLIT-like 2 monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.2 mg/mL of rabbit anti-Human Vasorin/SLIT-like 2 monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4°C).
  • Quantity 5 Plates
    Regulatory Status RUO
    Research Discipline Growth and Development, Neuronal Cell Markers
    Storage Requirements Storage is content dependent.
    Target Vasorin/SLIT-like 2
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 ug/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20°C to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 1500 pg/mL is recommended.
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.