missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Vasorin/SLIT-like 2 Antibody Pair [HRP]
Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
Description
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Vasorin/SLIT-like 2.
Specifications
Specifications
| Assay Range | 23.4 pg/mL |
| Assay Sensitivity | 23.4-1500 pg/mL |
| Assay | Sandwich ELISA |
| Conjugate | HRP |
| Product Type | Antibody Pairs |
| Specificity | This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Vasorin/SLIT-like 2. |
| Target Species | Human |
| For Use With (Application) | Sandwich ELISA |
| Gene Alias | Protein slit-like 2, SLITL2, slit-like 2, slit-like 2 (Drosophila), vasorin |
| Gene ID (Entrez) | 114990 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction