missing translation for 'onlineSavingsMsg'
Learn More

Vasorin/SLIT-like 2 Antibody, Novus Biologicals™

Product Code. 18399923 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18399923 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18399923 Supplier Novus Biologicals Supplier No. NBP318471

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Vasorin/SLIT-like 2 Polyclonal antibody specifically detects Vasorin/SLIT-like 2 in Human samples. It is validated for Western Blot, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen Vasorin/SLIT-like 2
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:10000 - 1:25000, Immunoprecipitation 50-100 ug/mg lysate
Formulation Tris-citrate/phosphate pH 7 -8.
Gene Alias Protein slit-like 2, SLITL2, slit-like 2, slit-like 2 (Drosophila), vasorin
Host Species Rabbit
Immunogen Recognizes region between residues Between between 623 and 673..
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Growth and Development, Neuronal Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 114990
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.