missing translation for 'onlineSavingsMsg'
Learn More

Vasorin/SLIT-like 2 Antibody (010), Janelia Fluor™ 646 , Novus Biologicals™

Product Code. 18426359 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18426359 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18426359 Supplier Novus Biologicals Supplier No. NBP290245JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Vasorin/SLIT-like 2 Monoclonal antibody specifically detects Vasorin/SLIT-like 2 in Human samples. It is validated for ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Vasorin/SLIT-like 2
Applications ELISA
Classification Monoclonal
Clone 010
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias Protein slit-like 2, SLITL2, slit-like 2, slit-like 2 (Drosophila), vasorin
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Vasorin/SLIT-like 2 (Q6EMK4; Met1-Pro575).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Growth and Development, Neuronal Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 114990
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.