Learn More
Vasopressin, 98+%
Description
It is used as a peptide hormone that regulates the body’s retention of water. Vasopressin or antidiuretic hormone is a potent endogenous hormone which is responsible for regulating plasma osmolality and volume. It acts as a neurotransmitter in the brain to control circadian rhythm, thermoregulation, and adrenocorticotrophic hormone release (ACTH). The therapeutic use of vasopressin has become increasingly important in the critical care environment in the management of cranial diabetes insipidus, bleeding abnormalities, oesophageal variceal haemorrhage, asystolic cardiac arrest, and septic shock.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Soluble in water (Miscible).
Notes
Store in airtight containers at 2° to 8°. Store in cool, sealed container.
Specifications
Specifications
| Color | White |
| Quantity | 100 mg |
| Solubility Information | Soluble in water (Miscible). |
| Formula Weight | 1084.24 |
| Percent Purity | ≥98% |
| Physical Form | Lyophilized Powder |
| Chemical Name or Material | Vasopressin |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.