missing translation for 'onlineSavingsMsg'
Learn More

Vasopressin, 98+%

Product Code. 15473859
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Unit Size:
100mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15473859 25 mg 25mg
15463859 5 mg 5mg
15483859 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15473859 Supplier Thermo Scientific Alfa Aesar Supplier No. J61248.MB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 11000-17-2 | C46H65N15O12S2

It is used as a peptide hormone that regulates the body’s retention of water. Vasopressin or antidiuretic hormone is a potent endogenous hormone which is responsible for regulating plasma osmolality and volume. It acts as a neurotransmitter in the brain to control circadian rhythm, thermoregulation, and adrenocorticotrophic hormone release (ACTH). The therapeutic use of vasopressin has become increasingly important in the critical care environment in the management of cranial diabetes insipidus, bleeding abnormalities, oesophageal variceal haemorrhage, asystolic cardiac arrest, and septic shock.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water (Miscible).

Notes
Store in airtight containers at 2° to 8°. Store in cool, sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11000-17-2
Molecular Formula C46H65N15O12S2
MDL Number MFCD00076738
Synonym Cys-Tyr-Phe-Gln-Asn-Cys-Pro-Arg-Gly-NH2; AVP peptide

Specifications

Color White
Quantity 25 mg
Solubility Information Soluble in water (Miscible).
Formula Weight 1084.24
Percent Purity ≥98%
Physical Form Lyophilized Powder
Chemical Name or Material Vasopressin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.