missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VASN Polyclonal Antibody

VASN Polyclonal Antibody
Brand: Invitrogen PA5110564
This item is not returnable.
View return policy
Description
VASN Polyclonal Antibody for Western Blot
Specifications
| VASN | |
| Polyclonal | |
| Unconjugated | |
| VASN | |
| 2610528G05Rik; Atia; Protein slit-like 2; Slitl2; Slit-like 2; UNQ314/PRO357/PRO1282; VASN; Vasorin | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 246154, 679921 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.2 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9CZT5 | |
| VASN | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 420-575 of human VASN (NP_612449.2). | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction