missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: GALGMSRRED LMYKPPAFRT LSELVLDFEA AYGRCWHVLK KVKLGQSVSH DPHSVEQIEW KHSVLDVERL GRDDFRKELE RHARDMRLKI GKGTGPPSPT KDRKKDVSSP QRAQSSPHRR NSRSERRPSG DKKTSEPKAM PD Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
VASH1 is a protein that possesses actin binding activity and metallocarboxypeptidase activity. It is involved in several processes including negative regulation of angiogenesis, negative regulation of blood vessel endothelial cell migration, and proteolysis. VASH1 acts upstream of or within processes such as negative regulation of endothelial cell migration, negative regulation of endothelial cell proliferation, and negative regulation of lymphangiogenesis. It is located in the apical part of the cell, endoplasmic reticulum, and extracellular space. VASH1 is implicated in liver cirrhosis and portal hypertension, and is a biomarker of liver cirrhosis. Diseases associated with VASH1 include Portal Hypertension and Myasthenic Syndrome, Congenital, 14.
Specifications
Specifications
| Antigen | VASH1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | VASH1 |
| Gene Accession No. | Q7L8A9 |
| Gene Alias | AI834978; D930046M13Rik; G630009D10Rik; KIAA1036; RGD1564082; TTCP 1; Tubulin carboxypeptidase 1; Tubulinyl-Tyr carboxypeptidase 1; Tyrosine carboxypeptidase 1; VASH; VASH1; vasohibin 1; vasohibin-1 |
| Gene Symbols | VASH1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?