missing translation for 'onlineSavingsMsg'
Learn More

VASA Antibody, Novus Biologicals™

Product Code. 18130972 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
Unit Size:
0.1mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18130972 0.025 mg 25µg
18797933 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18130972 Supplier Novus Biologicals Supplier No. NBP224558SS

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 2 publications

VASA Polyclonal specifically detects VASA in Human, Mouse, Rat, Porcine, Bovine, Canine, Equine, Primate, Xenopus, Zebrafish samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VASA
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 34183774), Immunohistochemistry-Paraffin 5 ug/ml
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. AF262962.1
Gene Alias DEAD (Asp-Glu-Ala-Asp) box polypeptide 4, DEAD box protein 4, EC 3.6.1, EC 3.6.4.13, MGC111074, probable ATP-dependent RNA helicase DDX4, Vasa homolog, VASADEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 4
Gene Symbols DDX4
Host Species Rabbit
Immunogen A portion of amino acids 700-724 of DDX4 was used as the immunogen.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54514
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse, cow, Xenopus, rat, horse, orangutan, zebrafish, and pig.
Target Species Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Primate, Xenopus species, Zebrafish
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.