missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ VAS 2870

Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Descrizione
VAS 2870 is a NADPH oxidase (Nox) inhibitor. Inhibits PMA-stimulated oxidative burst in HL-60 cells (IC50 = 2 μM). Inhibits oxidized-LDL-mediated, but not basal, ROS production in HUVECs in vitro. Also inhibits PDGF-mediated Nox activation and ROS formation in vascular smooth muscle cells (VSMCs) and abolishes PDGF-dependent migration of VSMCs, while having no effect on proliferation. Enhances TGF-β-induced apoptosis of liver tumor cells. Also inhibits platelet aggregation through a NOX-independent mechanism.
Specifica
Specifica
| Chemical Name or Material | 7-(2-Benzoxazolylthio)-3-(phenylmethyl)-3H-1,2,3-triazolo[4,5-d]pyrimidine |
| CAS | 722456-31-7 |
| Quantity | 25 mg |
| Molecular Formula | C18H12N6OS |
| Formula Weight | 360.39 |
| Purity | 98% |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto