missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VARS2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£157.00 - £364.00
Specifications
| Antigen | VARS2 |
|---|---|
| Dilution | Western Blot 1:200-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
VARS2 Polyclonal antibody specifically detects VARS2 in Human samples. It is validated for Western BlotSpecifications
| VARS2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| DKFZP434L1435, EC 6.1.1, EC 6.1.1.9, Em:AB023048.4, KIAA1885MGC138259, MGC142165, valine tRNA ligase 2, mitochondrial (putative), Valine--tRNA ligase, valRS, valyl-tRNA synthetase 2, mitochondrial (putative), valyl-tRNA synthetase 2-like, valyl-tRNA synthetase like, valyl-tRNA synthetase, mitochondrial, Valyl-tRNA synthetase-like, VARS2L, VARSL | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 864-1063 of human VARS2 (NP_065175.4). LWQRLPPRPGCPPAPSISVAPYPSACSLEHWRQPELERRFSRVQEVVQVLRALRATYQLTKARPRVLLQSSEPGDQGLFEAFLEPLGTLGYCGAVGLLPPGAAAPSGWAQAPLSDTAQVYMELQGLVDPQIQLPLLAARRYKLQKQLDSLTARTPSEGEAGTQRQQKLSSLQLELSKLDKAASHLRQLMDEPPAPGSPEL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:200-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 57176 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title