Learn More
Invitrogen™ VARS Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113815
Description
Immunogen sequence: AQRLRERRAA SGYPVKVPLE VQEADEAKLQ QTEAELRKVD EAIALFQKML For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 86%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 93%.
Aminoacyl-tRNA synthetases catalyze the aminoacylation of tRNA by their cognate amino acid. Because of their central role in linking amino acids with nucleotide triplets contained in tRNAs, aminoacyl-tRNA synthetases are thought to be among the first proteins that appeared in evolution. This protein belongs to the class-I aminoacyl-tRNA synthetase family and is located in the class III region of the major histocompatibility complex.
Specifications
| VARS | |
| Polyclonal | |
| Unconjugated | |
| VARS1 | |
| Bat6; D17H6S56E; EC 6.1.1.9; G7A; Protein G7a; TrsVal; valine tRNA ligase 1, cytoplasmic; valine--tRNA ligase; ValRS; valyl trna synthetase; valyl-tRNA synthetase; valyl-tRNA synthetase 2; VARS; VARS1; VARS2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 7407 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P26640 | |
| VARS1 | |
| Synthetic peptide directed towards the middle region of human VARS. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.