Learn More
Thermo Scientific™ AlphaScreen Upgrade for Varioskan™ LUX multimode microplate reader
Various accessories for use with the Varioskan™ LUX Multimode Microplate Reader
Brand:  Thermo Scientific™ VLUPA003
Product Code. 15390777
  • £7180.00 / Pack of 1
Estimated Shipment: 31-07-2026
Add to basket
Description
Description
Designed for bioscience researchers with a wide variety of needs, the Thermo Scientific™ Varioskan™ LUX multimode microplate reader comes equipped with a flexible range of measurement technologies including Absorbance, Fluorescence Intensity, Luminescence, AlphaScreen, and Time-Resolved Fluorescence. The instrument simplifies measurement setup with the automatic dynamic range selection, and its smart safety controls ease workflow and help avoid experimental errors. Varioskan LUX raises the bar for reliability and ease, with technology that works in researchers’ best interest.
The Varioskan LUX multimode reader has received HTRF™ certification from Cisbio Bioassays and DLReady™ validation from Promega Corporation.
Varioskan LUX is paired with the powerful fourth generation SkanIt Software, providing excellent usability and flexibility.
  • License free—Each user can install the software on his/her computer
  • Intuitive and flexible interface for multiple users and assays
  • Innovative Virtual Pipetting ToolTM makes it easy to define samples to plate layout
  • Measurement data is continuously saved to the database, so there’s no data lost due to unexpected interruptions such as power outages or accidental aborting
  • Simplified data processing and export
  • Export data in a single mouse click, or have it emailed automatically at the end of the run
  • Several file formats for exporting results: *.xlsx, *.pdf, *.xml, and *.txt
The Varioskan LUX is modular and upgradeable, so you can build the system that best suits your needs. For more information, contact a sales representative.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
AlphaScreen Upgrade
Varioskan LUX Microplate Reader