missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Varicella Zoster Virus Glycoprotein E Antibody (276), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-31978-20ug
This item is not returnable.
View return policy
Description
Varicella Zoster Virus Glycoprotein E Monoclonal antibody specifically detects Varicella Zoster Virus Glycoprotein E in Virus samples. It is validated for ELISA,Western Blot
Specifications
| Varicella Zoster Virus Glycoprotein E | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
| ELISA, Western Blot | |
| 276 | |
| Western Blot 0.1-1 μg/mL, ELISA 0.1-0.2 μg/mL | |
| Envelope glycoprotein E, GE, ORF68 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Varicella Zoster Virus Glycoprotein E (Uniprot#: Q9J3M8; Met1-Tyr538) | |
| 20 μg | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction