missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Varicella Zoster Virus Glycoprotein E Antibody (144), Novus Biologicals™
Rabbit Monoclonal Antibody
£164.00 - £343.00
Specifications
| Antigen | Varicella Zoster Virus Glycoprotein E |
|---|---|
| Clone | 144 |
| Dilution | Western Blot 0.1-1 μg/mL, ELISA 0.1-0.2 μg/mL |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30114510
|
Novus Biologicals
NBP3-31977-100ug |
100 μg |
£343.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30116296
|
Novus Biologicals
NBP3-31977-20ug |
20 μg |
£164.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Varicella Zoster Virus Glycoprotein E Monoclonal antibody specifically detects Varicella Zoster Virus Glycoprotein E in Virus samples. It is validated for ELISA,Western BlotSpecifications
| Varicella Zoster Virus Glycoprotein E | |
| Western Blot 0.1-1 μg/mL, ELISA 0.1-0.2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Varicella Zoster Virus Glycoprotein E (Uniprot#: Q9J3M8; Met1-Tyr538) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 144 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Virus | |
| Envelope glycoprotein E, GE, ORF68 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title