missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Varicella Zoster Virus Glycoprotein E Antibody (09) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

£167.00 - £360.00

Specifications

Antigen Varicella Zoster Virus Glycoprotein E
Clone 09
Dilution Western Blot 0.2-1 μg/mL, ELISA 0.5-1 μg/mL, Sandwich ELISA Detection 0.1-1 μg/mL
Applications ELISA, Western Blot, Sandwich ELISA Detection
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30028260
View Documents
Novus Biologicals™
NBP3-26914-20ug
20 μg
£167.00
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30025020
View Documents
Novus Biologicals™
NBP3-26914-100ug
100 μg
£360.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Varicella Zoster Virus Glycoprotein E Monoclonal antibody specifically detects Varicella Zoster Virus Glycoprotein E in Virus samples. It is validated for ELISA,Western Blot,Sandwich ELISA Detection
Specifications

Especificaciones

Varicella Zoster Virus Glycoprotein E
Western Blot 0.2-1 μg/mL, ELISA 0.5-1 μg/mL, Sandwich ELISA Detection 0.1-1 μg/mL
Monoclonal
Purified
RUO
0.2 μm filtered solution in PBS
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Varicella Zoster Virus Glycoprotein E (Uniprot#: Q9J3M8; Met1-Tyr538).
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
09
ELISA, Western Blot, Sandwich ELISA Detection
Unconjugated
Mouse
Virus
Envelope glycoprotein E, GE, ORF68
IgG
Protein A purified
Videos
SDS
Documentos

Documentos

Product Certifications
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado