missing translation for 'onlineSavingsMsg'
Learn More

Berthold Technologies Variable JET Injector (#3)

Product Code. 31777429 Shop All Berthold Technologies Products
Change view
Click to view available options
Volume (Metric) Dispense:
10 to 100 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Dispense Unit Size
31777429 10 to 100 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31777429 Supplier Berthold Technologies Supplier No. 7037903
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Injector for reagent dispensing in compatible luminometers with flexible installation options.

  • Centro supports up to three JET injectors with variable volumes in pre-measurement or measurement positions
  • Ultra-fast injection minimizes delay between injection and measurement
  • Suitable for measuring fast reactions such as flash luminescence
  • Uses Teflon bellows for fast and accurate injections
  • Provides efficient mixing and long service life
  • Delivers accurate injection of small and large volumes with >98% precision across the volume range
  • Designed for low maintenance due to frictionless operation
  • Compatible with cell suspensions due to low shear forces and appropriate materials

Specifications

Injection Type JET Injectors
Volume (Metric) Dispense 10 to 100 μL
Color Gray
For Use With (Application) Luminescence Applications
Voltage 24 VDC
For Use With (Equipment) Centro LB 963 Microplate Luminometer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.