missing translation for 'onlineSavingsMsg'
Learn More

Scie Plas Vari-Gel Tank Mini System with 2-Gel Tray

Ideal for drying sequencing gels or multiple small format gels.

Brand:  Scie Plas SVG-SYS


Additional Details : Weight : 0.80000kg

Product Code. 15895541

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • Two different gel trays to accomodate varying sizes (00mm and 70mm)
  • Multiple comb slots
  • Colour coded combs
  • Buffer recirculation ports, may be connected to peristaltic pump
  • Coloured loading strips for easy well detection when loading
  • Compact tank to reduce buffer volume required to cover gel
  • UV transparent acrylic casting tray

Comb Specifications:

  • Power output connector: Shrouded, 4mm (diameter)
  • Comb thickness: 1, 1.5 or 2mm
  • Comb throughput: 10 to 30 samples
  • Comb slots: 12
  • Migration distance between comb slots: 2.0cm
Specifications

Specifications

Gel Unit
Two removable gel trays, 15-sample combs, aluminum casting gates, coloured loading strips
950 mL
75 to 150 V
70, 100 x 70 mm
48
11 x 25 x 7 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers