missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Varespladib, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
Varespladib (LY315920) is a potent and selective group IIA, secretory phospholipase A2 (sPLA2) inhibitor with an IC50 of 9 nM. Varespladib exhibits the significant inhibitory effect on sPLA2 activity in serum from various species including rat, rabbit, guinea pig and human with IC50s of 8.1 nM, 5.0 nM, 3.2 nM and 6.2 nM, respectively.
Specifications
Specifications
| Chemical Name or Material | Varespladib |
| CAS | 172732-68-2 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C21H20N2O5 |
| Quantity | 50 mg |
| Synonym | LY315920 |
| Solubility Information | DMSO : ≥ 32 mg/mL (84.12 mM) ∣H2O : < 0.1 mg/mL (insoluble) |
| SMILES | O=C(O)COC1=CC=CC2=C1C(C(C(N)=O)=O)=C(CC)N2CC3=CC=CC=C3 |
| Molecular Weight (g/mol) | 380.39 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction