missing translation for 'onlineSavingsMsg'
Learn More

Varenicline, MedChemExpress

Product Code. 16402838
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16402838

Brand: Medchem Express HY10019

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Varenicline (CP 526555) is a potent partial agonist for α4β2 nicotinic acetylcholine receptor (nAChR) with an EC50 value of 2.3 μM. Varenicline is a full agonist for α3β4 and α7 nAChRs with EC50 values of 55 μM and 18 μM, respectively. Varenicline is a nicotinic ligand based on the structure of cytosine, and has the potential for smoking cessation treatment.

Varenicline (CP 526555) is a potent partial agonist for α4β2 nicotinic acetylcholine receptor (nAChR) with an EC50 value of 2.3 μM. Varenicline is a full agonist for α3β4 and α7 nAChRs with EC50 values of 55 μM and 18 μM, respectively. Varenicline is a nicotinic ligand based on the structure of cytosine, and has the potential for smoking cessation treatment.

Specifications

Chemical Name or Material Varenicline
CAS 249296-44-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C13H13N3
Quantity 10 mg
Synonym CP 526555
Solubility Information DMSO : 25 mg/mL (118.34 mM; Need ultrasonic)
SMILES C12=C(C=C3N=CC=NC3=C2)C4CC1CNC4
Molecular Weight (g/mol) 211.26
Formula Weight 211.26
Percent Purity 99.98%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.