missing translation for 'onlineSavingsMsg'
Learn More

Vapreotide Acetate, TRC

Product Code. 30451102
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30451102 50 mg -
30456002 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30451102 Supplier Toronto Research Chemicals Supplier No. TRCV09780050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 116430-60-5
Molecular Formula C59H74N12O11S2
Molecular Weight (g/mol) 1191.42
Synonym D-phenylalanyl-L-cysteinyl-L-tyrosyl-D-tryptophyl-L-lysyl-L-valyl-L-cysteinyl-L-Tryptophanamide Cyclic (2→7)-disulfide 1,2-Dithia-5,8,11,14,17-pentaazacycloeicosane, Cyclic Peptide Deriv., 16: PN: US20020042374 PAGE: 10 Claimed Protein, 20: PN: US6268342 SEQID: 23 Claimed Protein, 3: PN: EP1118336 SEQID: 3 Claimed Protein, BMY 41606, Octastatin, RC 160, RC 160 (protein)
SMILES O=C(N[C@H](CC1=CNC2=C1C=CC=C2)C3=O)[C@@H](NC([C@@H](NC([C@H](N)CC4=CC=CC=C4)=O)CSSC[C@@H](C(N[C@H](C(N)=O)CC5=CNC6=C5C=CC=C6)=O)NC([C@H](C(C)C)NC([C@H](CCCCN)N3)=O)=O)=O)CC7=CC=C(O)C=C7.CC(O)=O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C57H70N12O9S2.C2H4O2/c1-32(2)49-57(78)68-48(55(76)64-44(50(60)71)26-35-28-61-41-16-8-6-14-38(35)41)31-80-79-30-47(67-51(72)40(59)24-33-12-4-3-5-13-33)56(77)65-45(25-34-19-21-37(70)22-20-34)53(74)66-46(27-36-29-62-42-17-9-7-15-39(36)42)54(75)63-43(52(73)69-49)18-10-11-23-58;1-2(3)4/h3-9,12-17,19-22,28-29,32,40,43-49,61-62,70H,10-11,18,23-27,30-31,58-59H2,1-2H3,(H2,60,71)(H,63,75)(H,64,76)(H,65,77)(H,66,74)(H,67,72)(H,68,78)(H,69,73);1H3,(H,3,4)/t40-,43+,44+,45+,46-,47+,48+,49+;/m1./s1
Formula Weight 1190.5041
Chemical Name or Material Vapreotide Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.