Learn More
Cole-Parmer™ VapLock™ Solvent Waste Kits for Bottles and Carboys
VapLock™ Solvent Waste Kits for Bottles and Carboys collect effluent from multiple sources while minimizing harmful solvent exhaust
Brand: Cole-Parmer™ WAK38WP-10
Additional Details : Weight : 0.91000kg
Description
Various cap sizes available to fit your solvent containerServices
None
Specifications
Solvent Safety | |
Solvent Waste Kit | |
Solvent-Compatible Containers |
Wako 38 mm | |
Polypropylene | |
1 x 1/2 in. NPT(F), 3 x 1/4 inNPT(F), 6 x 1/4 in.-28 UN(F)(F) |