Learn More
VAPB Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14477-1-AP-20UL
 View more versions of this product
Product Code. 16853134
  • £135.00 / 20µL
Description
The VAPB antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human VAPB. This antibody recognizes human, mouse, rat antigen. The VAPB antibody has been validated for the following applications: FC, IF, IHC, IP, WB, ELISA analysis.
Specifications
Specifications
VAPB
Polyclonal
Unconjugated
20 μL
9217
IgG
Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, Flow Cytometry
300 μg/mL
Rabbit
Primary
Human, Rat, Mouse