missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VAPA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15275-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is an integral component of Cajal bodies (also called coiled bodies). Cajal bodies are nuclear suborganelles of varying number and composition that are involved in the post-transcriptional modification of small nuclear and small nucleolar RNAs. The N-terminus of the coilin protein directs its self-oligomerization while the C-terminus influences the number of nuclear bodies assembled per cell. Differential methylation and phosphorylation of coilin likely influences its localization among nuclear bodies and the composition and assembly of Cajal bodies. This gene has pseudogenes on chromosome 4 and chromosome 14.Specifications
| VAPA | |
| Polyclonal | |
| Unconjugated | |
| VAPA | |
| 33 kDa VAMP associated protein, hVAP 33, VAMP A, VAMP associated protein A, VAP 33, VAP A, VAP33, VAPA | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 30960, 58857, 9218 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot | |
| 0.42 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9P0L0, Q9WV55, Q9Z270 | |
| Vapa | |
| VAPA Fusion Protein Ag7393 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction