Learn More
VAP-B Antibody (736904) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB58551JF585
Product Code. 30172814
  • £442.00 / 0.10mL
Description
VAP-B Monoclonal antibody specifically detects VAP-B in Human samples. It is validated for Immunohistochemistry,Western Blot,Flow Cytometry (Intracellular Staining),CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
VAP-B
Monoclonal
Janelia Fluor 585
ALS8VAMP-B/VAMP-C, VAMP (vesicle-associated membrane protein)-associated protein B and C, VAMP-associated 33 kDa protein, VAMP-associated protein B/C, VAMP-B, VAP-B, VAP-B/VAP-C, VAPC, VAP-C, vesicle-associated membrane protein-associated protein B/C
E. coli-derived recombinant human VAP-B, Ala2-Pro132, Accession # O95292
RUO
Primary
Human
Liquid
Immunohistochemistry, Western Blot, Flow Cytometry (Intracellular Staining), CyTOF
736904
50mM Sodium Borate
Mouse
0.1 mL
Neuroscience
9217
Store at 4°C in the dark.
IgG1