missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VAP-B Antibody (736904), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB58551CL1
This item is not returnable.
View return policy
Description
VAP-B Monoclonal antibody specifically detects VAP-B in Human samples. It is validated for Immunohistochemistry,Western Blot,Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| VAP-B | |
| Monoclonal | |
| CoraFluor 1 | |
| ALS8VAMP-B/VAMP-C, VAMP (vesicle-associated membrane protein)-associated protein B and C, VAMP-associated 33 kDa protein, VAMP-associated protein B/C, VAMP-B, VAP-B, VAP-B/VAP-C, VAPC, VAP-C, vesicle-associated membrane protein-associated protein B/C | |
| E. coli-derived recombinant human VAP-B, Ala2-Pro132, Accession # O95292 | |
| 0.1 mL | |
| Neuroscience | |
| 9217 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
| 736904 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction