missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VAP-B Antibody (208), mFluor Violet 450 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89626MFV450
This item is not returnable.
View return policy
Description
VAP-B Monoclonal antibody specifically detects VAP-B in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation (Negative)
Specifications
| VAP-B | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 208 | |
| Western Blot, ELISA, Immunoprecipitation (Negative) | |
| ALS8VAMP-B/VAMP-C, VAMP (vesicle-associated membrane protein)-associated protein B and C, VAMP-associated 33 kDa protein, VAMP-associated protein B/C, VAMP-B, VAP-B, VAP-B/VAP-C, VAPC, VAP-C, vesicle-associated membrane protein-associated protein B/C | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human VAP-B (Uniprot#: O95292-1; Met 1-Pro 132). | |
| 0.1 mL | |
| Neuroscience | |
| 9217 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction