missing translation for 'onlineSavingsMsg'
Learn More

VAP-A Antibody, HRP, Novus Biologicals™

Product Code. 18421062 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18421062 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18421062 Supplier Novus Biologicals Supplier No. NBP232249H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

VAP-A Polyclonal antibody specifically detects VAP-A in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen VAP-A
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate HRP
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias hVAP-33, MGC3745,33 kDa VAMP-associated protein, VAMP (vesicle-associated membrane protein)-associated protein A, 33kDa, VAMP-A, VAP-33, VAP33VAMP (vesicle-associated membrane protein)-associated protein A (33kD), VAP-AVAMP-associated protein A, vesicle-associated membrane protein-associated protein A
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 125 to 175 of human 33 kDa VAMP-Associated Protein using the numbering given in entry NP_919415.2 (GeneID 9218).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9218
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.