Learn More
Invitrogen™ VAP-1 Monoclonal Antibody (TK8-14), eBioscience™

Mouse Monoclonal Antibody
Brand: Invitrogen™ BMS1033
Description
Description: VAP-1 (TK8-14) is a mouse monoclonal antibody raised against affinity purifiedVAP-1 from tonsil stroma of human origin. Lymphocyte binding to vascular endothelium is a prerequisite for the move-ment of immune cells from the blood into lymphoid tissues and into sites ofinflammation. Under inflammatory conditions, cell surface expression ofVAP-1 (vascular adhesion protein-1) which is an endothelial sialoglycoprotein,is induced. VAP-1 is a type II transmembrane protein of 84.6 kDa with a singletransmembrane domain and N and O-glycosylation sites in the extracellulardomain. In vivo, VAP-1 exists predominantly as a 180 kDa homodimer andfunctions both as an enzyme (monoamine oxidase) and an adhesion moleculefor lymphocytes. Applications Tested: ELISA, Flow Cytometry, Immunofluorescence, Western Blotting.
Copper amine oxidases catalyze the oxidative conversion of amines to aldehydes in the presence of copper and quinone cofactor. The product is a major protein on the adipocyte plasma membrane. It has adhesive properties and also has functional monoamine oxidase activity. A pseudogene for this gene has been discribed and is located approximately 9-kb downstream.
Specifications
| VAP-1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.2-7.4 | |
| Q16853 | |
| AOC3 | |
| Rec. Human VAP-1. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| ELISA, Flow Cytometry, Immunohistochemistry, Western Blot | |
| TK8-14 | |
| Unconjugated | |
| AOC3 | |
| amine oxidase, copper containing 3; amine oxidase, copper containing 3 (vascular adhesion protein 1); Aoc3; Copper amine oxidase; HPAO; Membrane primary amine oxidase; placenta copper monamine oxidase; semicarbazide-sensitive amine oxidase; SSAO; VAP1; VAP-1; Vascular adhesion protein 1; VP97 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 8639 | |
| -20°C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.