missing translation for 'onlineSavingsMsg'
Learn More

VAP-1/AOC3, Rabbit anti-Human, Clone: TK8-14, Novus Biologicals™

Rabbit Monoclonal Antibody

£207.00 - £463.00

Specifications

Antigen VAP-1/AOC3
Clone TK8-14
Concentration 1 mg/mL
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18355011
View Documents
Novus Biologicals
NBP2-81043-0.025MG
0.025 mg
£207.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18347560
View Documents
Novus Biologicals
NBP2-81043-0.2MG
0.2 mg
£463.00
0.20mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

VAP-1/AOC3 Monoclonal specifically detects VAP-1/AOC3 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications

Specifications

VAP-1/AOC3
1 mg/mL
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation
Unconjugated
Rabbit
Human
amine oxidase, copper containing 3 (vascular adhesion protein 1), Copper amine oxidase, EC 1.4.3, HPAOSSAO, Semicarbazide-sensitive amine oxidase, VAP1EC 1.4.3.21, VAP-1membrane primary amine oxidase, Vascular adhesion protein 1
AOC3
IgG κ
Protein A purified
Recognises human Vascular Adhesion Protein-1 (VAP-1/AOC3), a glycosylated homodimeric membrane protein consisting of two 90 kDa subunits connected by disulfide bonds. Epitope recognized by TK8-18 is present both in dimeric and monomeric forms of VAP-1/AOC3. Elevated soluble VAP-1/AOC3 serum- levels have been described in several inflammatory- diseases as well as colorectal cancer. TK8-14 is a function-blocking mAb.
TK8-14
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation
Monoclonal
Purified
RUO
PBS
8639
Affinity purified VAP-1/AOC3 from tonsil stroma of human origin.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.