missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
VAP-1/AOC3 Monoclonal antibody specifically detects VAP-1/AOC3 in Human samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antigen | VAP-1/AOC3 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | TK8-14 |
| Conjugate | mFluor Violet 610 SE |
| Dilution | Flow Cytometry |
| Formulation | 50mM Sodium Borate |
| Gene Alias | amine oxidase, copper containing 3 (vascular adhesion protein 1), Copper amine oxidase, EC 1.4.3, HPAOSSAO, Semicarbazide-sensitive amine oxidase, VAP1EC 1.4.3.21, VAP-1membrane primary amine oxidase, Vascular adhesion protein 1 |
| Host Species | Rabbit |
| Immunogen | Affinity purified VAP-1/AOC3 from tonsil stroma of human origin. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?