Learn More
Thermo Scientific™ Vanquish™ Split Sampler Sample Loops
Product Code. 17857828 Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
Vanquish Split Sampler HT and FT
Vanquish Split Sampler NT
Sample Capacity:
10 μL
25 μL
100 μL
Biocompatible Version:
Yes
Unit Size:
Each
Product Code. 17857828
Brand: Thermo Scientific™ 6252.1940
This item requires a price update prior to being ordered.
Request a Price for this item here.
Thermo Scientific™ Vanquish™ Split Sampler Sample Loops come in a variety of sizes to fit your application requirements.
Automate sample injection into your Thermo Scientific™ Vanquish™ HPLC or UHPLC system with Thermo Scientific™ Vanquish™ Split Sampler Sample Loops. These sample loops are available in 10, 25, 100, 250, 1000 μL sizes to meet your application requirements. Thermo Scientific™ Viper™ Fittings are included with our sample loops to make them easy to install with leak-free connections.
Vanquish Split Sampler Sample Loops use Viper Fittings, which have a ferrule-free tightening design, for virtually zero-dead-volume connections.
Specifications
Type Sample Loop
Sample Capacity 25 μL
Biocompatible Version Yes
For Use With (Equipment) Vanquish Split Sampler NT