Learn More
Thermo Scientific™ Vanquish™ Split Sampler Sample Loops
Product Code. 16371298 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Vanquish Split Sampler HT and FT
Vanquish Split Sampler NT
Sample Capacity:
10 μL
25 μL
100 μL
Biocompatible Version:
Yes
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Sample Capacity Biocompatible Version unitSize
16371298 Vanquish Split Sampler HT and FT 10 μL Yes Each
17857828 Vanquish Split Sampler NT 25 μL Yes -
17867828 Vanquish Split Sampler NT 100 μL Yes Each
17877828 Vanquish Split Sampler NT 10 μL Yes -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16371298 Supplier Thermo Scientific™ Supplier No. 6850.1915
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Thermo Scientific™ Vanquish™ Split Sampler Sample Loops come in a variety of sizes to fit your application requirements..
Automate sample injection into your Thermo Scientific™ Vanquish™ HPLC or UHPLC system with Thermo Scientific™ Vanquish™ Split Sampler Sample Loops. These sample loops are available in 10, 25, 100, 250, 1000 μL sizes to meet your application requirements. Thermo Scientific™ Viper™ Fittings are included with our sample loops to make them easy to install with leak-free connections.
Vanquish Split Sampler Sample Loops use Viper Fittings, which have a ferrule-free tightening design, for virtually zero-dead-volume connections.
Specifications
Type Sample Loop
Sample Capacity 10 μL
Biocompatible Version Yes
For Use With (Equipment) Vanquish Split Sampler HT and FT