Learn More
Thermo Scientific™ Vanquish™ Flow Cells for Vanquish™ Diode Array Detector FG
Product Code. 17565446 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Standard Biocompatible Flow Cell
Unit Size:
Each
Product Code. Type Unit Size
17565446 Standard Biocompatible Flow Cell Each
1 options
Product Code. 17565446 Supplier Thermo Scientific™ Supplier No. 6083.0540
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Choose from a wide portfolio of flow cells for the Thermo Scientific™ Vanquish™ Diode Array Detector (DAD) FG to match your application-related requirements.
Choose from a wide portfolio of Thermo Scientific™ Vanquish™ Flow Cells for the Thermo Scientific™ Vanquish™ Diode Array Detector FG. There's a flow cell to match your application-related requirements including low-dispersion for UHPLC separations, high-pressure resistance for LC-MS or fractionation, and compatibility with the analysis of biomolecules.
Great flow cells for a wide range of applications
12 MPa (120 bar, 1740 psi) flow cell pressure limit:
  • Standard flow cell
  • Semi-analytical flow cell
  • Semi-micro flow cells (biocompatible and SST version available)
5 MPa (50 bar, 720 psi) flow cell pressure limit:
  • Standard biocompatible flow cell
Specifications
For Use With (Equipment) Vanquish Diode Array Detector FG
Material PEEK
Type Standard Biocompatible Flow Cell
Volume (Metric) 13 μL
Path Length 10 mm
Product Line Vanquish