missing translation for 'onlineSavingsMsg'
Learn More

Vanoxerine dihydrochloride, MedChemExpress

Product Code. 16465558
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16465558 10 mg 10mg
16441727 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16465558 Supplier Medchem Express Supplier No. HY13217
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Vanoxerine dihydrochloride (GBR-12909 dihydrochloride) is a competitive, potent, and highly selective dopamine reuptake inhibitor (Ki=1 nM). Vanoxerine dihydrochloride (GBR-12909 dihydrochloride) binds to the target site on the dopamine transporter (DAT).

Vanoxerine dihydrochloride (GBR-12909 dihydrochloride) is a competitive, potent, and highly selective dopamine reuptake inhibitor (Ki=1 nM). Vanoxerine dihydrochloride (GBR-12909 dihydrochloride) binds to the target site on the dopamine transporter (DAT).

Specifications

Chemical Name or Material Vanoxerine dihydrochloride
CAS 67469-78-7
Color White
Purity Grade Notes Research
Molecular Formula C28H34Cl2F2N2O
Quantity 10 mg
Synonym GBR-12909 dihydrochloride I893 dihydrochloride
Solubility Information DMSO : 9.4 mg/mL (17.96 mM; Need ultrasonic and warming) ∣H2O : 1 mg/mL (1.91 mM; ultrasonic and warming and heat to 60°C)
SMILES FC1=CC=C(C(C2=CC=C(F)C=C2)OCCN3CCN(CCCC4=CC=CC=C4)CC3)C=C1.[2HCl]
Molecular Weight (g/mol) 523.49
Formula Weight 523.49
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.