missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vanin-1/VNN1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98364-100ul
This item is not returnable.
View return policy
Description
Vanin-1/VNN1 Polyclonal antibody specifically detects Vanin-1/VNN1 in Mouse samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
Specifications
| Vanin-1/VNN1 | |
| Polyclonal | |
| ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin 1:500-1:2000 | |
| EC 3.5.1, EC 3.5.1.92, HDLCQ8, MGC116930, MGC116931, MGC116932, pantetheinase, Pantetheine hydrolase, Tiff66MGC116933, vanin 1, vanin-1, vannin 1, Vascular non-inflammatory molecule 1 | |
| Produced in rabbits immunized with purified, recombinant Mouse Vanin-1/VNN1 (Uniprot#: Q9Z0K8; Met1-Ser487) | |
| 100 μg | |
| Cancer, Cardiovascular Biology, Cell Biology, Endocrinology, Signal Transduction, Stem Cells | |
| 8876 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction