missing translation for 'onlineSavingsMsg'
Learn More

Vanilpyruvic acid, MedChemExpress

Product Code. 16423358
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16423358 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16423358 Supplier Medchem Express Supplier No. HY101416

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Vanilpyruvic acid is a catecholamine metabolite and precursor to vanillactic acid.

Vanilpyruvic acid is a catecholamine metabolite and precursor to vanillactic acid.

Specifications

Chemical Name or Material Vanilpyruvic acid
CAS 1081-71-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C10H10O5
Quantity 10 mg
Synonym Vanylpyruvic acid
Solubility Information DMSO : ≥ 27 mg/mL (128.46 mM)
SMILES O=C(O)C(CC1=CC=C(O)C(OC)=C1)=O
Molecular Weight (g/mol) 210.18
Formula Weight 210.18
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.