missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vanillyl Butyl Ether, TRC
Click to view available options
Quantity:
100 g
Unit Size:
100g
Specifications
Specifications
| Chemical Name or Material | Vanillyl Butyl Ether |
| CAS | 82654-98-6 |
| Molecular Formula | C12 H18 O3 |
| Quantity | 100 g |
| Synonym | 4-(Butoxymethyl)-2-methoxyphenol, Hotact VBE |
| InChI Formula | InChI=1S/C12H18O3/c1-3-4-7-15-9-10-5-6-11(13)12(8-10)14-2/h5-6,8,13H,3-4,7,9H2,1-2H3 |
| SMILES | CCCCOCc1ccc(O)c(OC)c1 |
| IUPAC Name | 4-(butoxymethyl)-2-methoxyphenol |
| Molecular Weight (g/mol) | 210.27 |
| Formula Weight | 210.1256 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction