missing translation for 'onlineSavingsMsg'
Learn More

Vanillin, TRC

Product Code. 30012189
Change view
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012189 100 g 100g
30011812 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30012189 Supplier Toronto Research Chemicals Supplier No. TRCV097500100G
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-33-5
Molecular Formula C8 H8 O3
Molecular Weight (g/mol) 152.15
Synonym Vanillin, Benzaldehyde, 4-hydroxy-3-methoxy-, Vanillin (8CI), 2-Methoxy-4-formylphenol, 3-Methoxy-4-hydroxybenzaldehyde, 4-Formyl-2-methoxyphenol, 4-Hydroxy-3-methoxy-benzyldehyde, 4-Hydroxy-3-methoxybenzaldehyde, 4-Hydroxy-5-methoxybenzaldehyde, 4-Hydroxy-m-anisaldehyde, H 0264, Lioxin, NPLC 0145, NSC 15351, NSC 403658, NSC 48383, Rhovanil, Vanillaldehyde, Vanillic aldehyde, Vanillum, m-Methoxy-p-hydroxybenzaldehyde, p-Hydroxy-m-methoxybenzaldehyde, p-Vanillin
IUPAC Name 4-hydroxy-3-methoxybenzaldehyde
SMILES COc1cc(C=O)ccc1O

Specifications

Quantity 100 g
InChI Formula InChI=1S/C8H8O3/c1-11-8-4-6(5-9)2-3-7(8)10/h2-5,10H,1H3
Formula Weight 152.0473
Percent Purity >95%
Chemical Name or Material Vanillin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.