missing translation for 'onlineSavingsMsg'
Learn More

Vanillin, Mikromol™

Product Code. 30050014
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30050014 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30050014 Supplier Mikromol Supplier No. MM1447.000250
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Vanillin
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Vanillin
CAS 121-33-5
Analyte or Component Names Vanillin
Shipping Condition Room Temperature
Molecular Formula C8 H8 O3
Certifications/Compliance ISO 17034
Synonym Vanillin, Benzaldehyde, 4-hydroxy-3-methoxy-, Vanillin (8CI), 2-Methoxy-4-formylphenol, 3-Methoxy-4-hydroxybenzaldehyde, 4-Formyl-2-methoxyphenol, 4-Hydroxy-3-methoxy-benzyldehyde, 4-Hydroxy-3-methoxybenzaldehyde, 4-Hydroxy-5-methoxybenzaldehyde, 4-Hydroxy-m-anisaldehyde, H 0264, Lioxin, NPLC 0145, NSC 15351, NSC 403658, NSC 48383, Rhovanil, Vanillaldehyde, Vanillic aldehyde, Vanillum, m-Methoxy-p-hydroxybenzaldehyde, p-Hydroxy-m-methoxybenzaldehyde, p-Vanillin
InChI Formula InChI=1S/C8H8O3/c1-11-8-4-6(5-9)2-3-7(8)10/h2-5,10H,1H3
SMILES COc1cc(C=O)ccc1O
IUPAC Name 4-hydroxy-3-methoxybenzaldehyde
Molecular Weight (g/mol) 152.15
Quantity 250 mg
Formula Weight 152.0473 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.