missing translation for 'onlineSavingsMsg'
Learn More

Vanillin 1000 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30322929
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30322929 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30322929 Supplier Dr. Ehrenstorfer Supplier No. DREA17900580AL1000
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Vanillin
CAS 121-33-5
Analyte or Component Names Vanillin
Shipping Condition Room Temperature
Molecular Formula C8 H8 O3
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym Vanillin, Benzaldehyde, 4-hydroxy-3-methoxy-, Vanillin (8CI), 2-Methoxy-4-formylphenol, 3-Methoxy-4-hydroxybenzaldehyde, 4-Formyl-2-methoxyphenol, 4-Hydroxy-3-methoxy-benzyldehyde, 4-Hydroxy-3-methoxybenzaldehyde, 4-Hydroxy-5-methoxybenzaldehyde, 4-Hydroxy-m-anisaldehyde, H 0264, Lioxin, NPLC 0145, NSC 15351, NSC 403658, NSC 48383, Rhovanil, Vanillaldehyde, Vanillic aldehyde, Vanillum, m-Methoxy-p-hydroxybenzaldehyde, p-Hydroxy-m-methoxybenzaldehyde, p-Vanillin
InChI Formula InChI=1S/C8H8O3/c1-11-8-4-6(5-9)2-3-7(8)10/h2-5,10H,1H3
SMILES COc1cc(C=O)ccc1O
IUPAC Name 4-hydroxy-3-methoxybenzaldehyde
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 152.15
Quantity 1 mL
Formula Weight 152.0473 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.