missing translation for 'onlineSavingsMsg'
Learn More

Vanillic acid, 98%

Product Code. p-7081893
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11449972

Brand: Thermo Scientific Alfa Aesar A12074.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 121-34-6 | C8H8O4 | 168.15 g/mol

Vanillic acid is used as a flavoring agent in food. It acts as an intermediate in the production of vanillin from ferulic acid. Further, it is used in wine and vinegar.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Vanillic acid is used as a flavoring agent in food. It acts as an intermediate in the production of vanillin from ferulic acid. Further, it is used in wine and vinegar.

Solubility
Soluble in water, alcohol and ether.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-34-6
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.15
MDL Number MFCD00002551
InChI Key WKOLLVMJNQIZCI-UHFFFAOYSA-N
Synonym vanillic acid, acide vanillique, p-vanillic acid, benzoic acid, 4-hydroxy-3-methoxy, 3-methoxy-4-hydroxybenzoic acid, vanillate, m-anisic acid, 4-hydroxy, protocatechuic acid, 3-methyl ester, 4-hydroxy-3-methoxy-benzoic acid, unii-gm8q3jm2y8
PubChem CID 8468
ChEBI CHEBI:30816
IUPAC Name 4-hydroxy-3-methoxybenzoic acid
SMILES COC1=CC(=CC=C1O)C(O)=O

Specifications

Melting Point 210°C to 218°C
Assay Percent Range 99%
Beilstein 2208364
Merck Index 14,9931
Solubility Information Soluble in water,alcohol and ether.
Formula Weight 168.15
Percent Purity 98%
Quantity 25 g
Chemical Name or Material Vanillic Acid
compliance-icons
Product Identifier
  • Vanillic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 4-hydroxy-3-methoxybenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.