missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VANGL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21492-1-AP-150UL
This item is not returnable.
View return policy
Description
Van Gogh is an alternate name for the Drosophila strabismus protein. Van Gogh like-2 (VANGL2) is a 521 amino acidprotein in vertebrates similar to strabismus 1 of Drosophila and a component of the frizzled-disheveled tissue polarity pathway. It interacts through it′s C-terminal region with the N-terminal half of DVL1, DVL2 and DVL3 and is a potent tumor suppressor. It ispredomitly involved in the control of early morphogenesis and patterning of both axial midline structures and the development of neural plate. It plays a crucial role in the regulation of planar cell polarity, particularly in the orientation of stereociliary bundles in the cochlea. VANGL2 is required for polarization and movement of myocardializing cells and seems to act via RHOA signaling to regulate this process. It is expressed in a wide range of tissues.Specifications
| VANGL2 | |
| Polyclonal | |
| Unconjugated | |
| VANGL2 | |
| KIAA1215, Loop tail protein 1 homolog, LPP1, LTAP, STB1, STBM, STBM1, Strabismus 1, Van Gogh like protein 2, Vang like protein 2, VANGL2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 289229, 57216, 93840 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P84889, Q91ZD4, Q9ULK5 | |
| Vangl2 | |
| VANGL2 Fusion Protein Ag15833 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction