Learn More
VANGL2 Mouse anti-Human, Unconjugated, Clone: 1B2C1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67273-1-IG-20UL
This item is not returnable.
View return policy
Description
The VANGL2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human VANGL2. This antibody recognizes human antigen. The VANGL2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| VANGL2 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot, ELISA | |
| 1B2C1 | |
| Unconjugated | |
| 20 μL | |
| 57216 | |
| IgG3 |