missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VANGL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14696-1-AP-150UL
This item is not returnable.
View return policy
Description
Vang-like 1 protein is a novel human homologue of Drosophila Van Gogh/Strabismus. The Vang-like 1 gene consists of at least 7 exons, and encodes a protein with four transmembrane domains and the C-terminal Ser/Thr-X-Val motif. Studies shows that the expression of the protein is significantly up-regulated in gastric cancer cell lines MKN28, MKN74, pancreatic cancer cell lines BxPC-3, PSN-1 and Hs766T. The protein may serve as an effector mediating the intestinal trefoil factor (ITF) healing response of the intestinal mucosa. The protein also serves as a diagnostic marker for hepatocellular carcinoma as well as a potential molecular target for development of novel therapeutic drugs. The pathological role of the protein has been identified in neural tube defects (NTD). Human Vang-like 1 gene maps to chromosome 1p11-p13.1.Specifications
| VANGL1 | |
| Polyclonal | |
| Unconjugated | |
| VANGL1 | |
| Loop tail protein 2 homolog, LPP2, STB2, STBM2, Strabismus 2, Van Gogh like protein 1, Vang like protein 1, VANGL1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 229658, 690366, 81839 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q80Z96, Q8TAA9 | |
| Vangl1 | |
| VANGL1 Fusion Protein Ag6306 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction