missing translation for 'onlineSavingsMsg'
Learn More

Vang-like Protein 2/VANGL2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-24704SS

 View more versions of this product

Product Code. 18170702

  • £167.00 / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Vang-like Protein 2/VANGL2 Polyclonal specifically detects Vang-like Protein 2/VANGL2 in Human, Mouse, Rat, Bovine, Canine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Vang-like Protein 2/VANGL2
Polyclonal
Unconjugated
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
VANGL2
A portion of amino acids 300-350 of human Van Gogh like-2 protein was used as the immunogen.
0.025 mg
Primary
100% homologous in human isoform CRA_a, CRA_b and CRA_c).
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG
Western Blot, Immunohistochemistry (Paraffin)
0.5 mg/mL
Western Blot 4-6 ug/ml, Immunohistochemistry-Paraffin
KIAA1215Loop-tail protein 1 homolog, loop-tail-associated protein, LPP1, LTAPVan Gogh-like protein 2, MGC119403, MGC119404, STB1Strabismus 1, STBM, STBM1, vang (van gogh, Drosophila)-like 2, vang, van gogh-like 2 (Drosophila), vang-like 2 (van gogh, Drosophila), vang-like protein 2
Rabbit
Protein G purified
RUO
57216
Human, Mouse, Rat, Bovine, Canine, Primate
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only