missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Vang-like Protein 1/VANGL1 Polyclonal specifically detects Vang-like Protein 1/VANGL1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Knockdown Validated.
Specifications
Specifications
| Antigen | Vang-like Protein 1/VANGL1 |
| Applications | Immunofluorescence, Immunocytochemistry, Immunofluorescence, KnockDown |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.4 ug/ml, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Knockdown Validated |
| Formulation | PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide |
| Gene Alias | KAI1 C-terminal interacting tetraspanin, Loop-tail protein 2 homolog, LPP2, MGC5338, STB2KITENIN, STBM2, Strabismus 2, Van Gogh-like protein 1, vang (van gogh, Drosophila)-like 1, vang, van gogh-like 1 (Drosophila), vang-like 1 (van gogh, Drosophila), vang-like protein 1 |
| Gene Symbols | VANGL1 |
| Host Species | Rabbit |
| Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:DTESTYSGYSYYSSHSKKSHRQGERTRERHKSPRNKDGRGSEKSVTIQPPTGEPLLGNDSTRTEEVQDDNWGETTTAITGTSEHSISQEDIARISKDMEDSVGLDCKRY |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?