missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Vandetanib
GREENER_CHOICE

Product Code. 18795836
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18795836 10 mg 10mg
18757046 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18795836 Supplier Tocris Bioscience™ Supplier No. 7497/10
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Potent VEGFR-2 inhibitor, also inhibits EGFR and RET oncoproteins activity

Vandetanib is a potent VEGFR-2 inhibitor (IC50 = 0.04 μM), and inhibitor of EGFR (IC50 = 0.5 μM). Vandetanib potently inhibits (IC50 = 100 nM) RET oncoprotein activity in human thyroid cancer. Vandetanib displays anti-angiogenic properties in human nasopharyngeal carcinoma in mice model. Vandetanib is orally available and effective in vivo.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(4-Bromo-2-fluorophenyl)-6-methoxy-7-[(1-methyl-4-piperidinyl)methoxy]-4-quinazolinamine
CAS 443913-73-3
Quantity 10 mg
Target VEGF Receptor Inhibitors
Molecular Formula C22H24BrFN4O2
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.