missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Vandetanib

Product Code. 17110242
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17110242 10 mg 10mg
17120242 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17110242 Supplier STEMCELL Technologies Supplier No. 73532
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Tyrosine kinase inhibitor; Inhibits VEGFR1, KDR, FLT-4, EGFR, FGFR, ABL, RET, and SRC

  • Vandetanib is a multi-kinase inhibitor targeting primarily receptor tyrosine kinases, such as vascular endothelial growth factor receptors (VEGFR1, KDR, and FLT4), epidermal growth factor receptor (EGFR), and fibroblast growth factor receptor (FGFR; Morabito et al.) It most potently inhibits KDR with an IC50 of 40 nM
  • It also inhibits non-receptor tyrosine kinases such as RET, ABL, and SRC as well as several serine/threonine kinases (Carlomagno et al.; Hennequin et al.; Kiselyov et al.; Levinson & Boxer)
  • Inhibits tumor growth in mouse xenograft models, including RET/PTC papillary thyroid carcinoma and lung carcinoma (Hennequin et al.; Carlomagno et al.)
  • Inhibits angiogenesis, cell growth, and metastasis in numerous cancer cell lines (Morabito et al.; Wedge et al.)

Specifications

CAS 443913-73-3
Synonym CH 331; ZD 6474; Zactima
Quantity 10 mg
Molecular Formula C22H24BrFN4O2
Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.